Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Coiled FEP, 3/8"" ID x 1/2"" OD; 2 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9582104
Encompass
Our retractable coil FEP tubing offers the chemical inertness of our standard FEP tubing combined with the flexibility of a coil. FEP tubing has excellent heat resistance, a low coefficient of friction, and won't deteriorate with age. Tubing withstands temperatures from -454 to 400¿F (-270 to 205¿C); and can be sterilized by autoclave, with ethylene oxide, or by using dry heat. Choose the size and length that best fits your application. Use tubing with compression fittings.
Catalog No. 50-634-315