Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, FEP, 1/16"" ID x 1/8"" OD; 25 ft
Catalog No. 50634320
Encompass
Excellent for the transfer of extreme temperature or highly corrosive materials. Also good for high-purity work. Chemically inert, nontoxic, heat resistant, with a low coefficient of friction. Doesnt deteriorate with age. Tough, but bends even at 454>F (270>C). 25FT per pack
Catalog No. 50-634-320 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640662
Excellent for the transfer of extreme temperature or highly corrosive materials. Also good for high-purity work. Chemically inert, nontoxic, heat resistant, with a low coefficient of friction. Doesnt deteriorate with age. Tough, but bends even at 454>F (270>C). 25FT per pack