Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, FEP, 1/2"" ID x 9/16"" OD; 25 ft
Catalog No. 50634328
Encompass
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Excellent for the transfer of extreme temperature or highly corrosive materials. Also good for high-purity work. Characteristics: Chemically inert, nontoxic, heat resistant, with a low coefficient of friction. Doesn¿t deteriorate with age. Tough, but bends even at -454¿F (-270¿C). Translucent. Certifications: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured in ISO 9001-2008 approved facility. Temperature range: -454 to 400¿F (-270 to 205¿C) Sterilize: Autoclave, ethylene oxide, or use dry heat.
Catalog No. 50-634-328 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640618
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Excellent for the transfer of extreme temperature or highly corrosive materials. Also good for high-purity work. Characteristics: Chemically inert, nontoxic, heat resistant, with a low coefficient of friction. Doesn¿t deteriorate with age. Tough, but bends even at -454¿F (-270¿C). Translucent. Certifications: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured in ISO 9001-2008 approved facility. Temperature range: -454 to 400¿F (-270 to 205¿C) Sterilize: Autoclave, ethylene oxide, or use dry heat.