Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, FEP-Lined Polyethylene, 3/8"" ID x 1/2"" OD; 100 ft
Catalog No. 50634295
Encompass
Applications: Ground water sampling and other sampling and monitoring uses that require the high purity and chemical inertness of FEP. Characteristics: Chemically inert, non-toxic FEP-liner with a durable polyethylene jacket. Rigid but bends at low temperatures. Translucent. Certification: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured according to GMP. Temperature range: -100 to 175¿F (-73 to 80¿C) Sterilize: With ethylene oxide.
Catalog No. 50-634-295 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0638504
Applications: Ground water sampling and other sampling and monitoring uses that require the high purity and chemical inertness of FEP. Characteristics: Chemically inert, non-toxic FEP-liner with a durable polyethylene jacket. Rigid but bends at low temperatures. Translucent. Certification: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured according to GMP. Temperature range: -100 to 175¿F (-73 to 80¿C) Sterilize: With ethylene oxide.