Learn More
AVANTOR MASTERFLEX (VWR LLC) Flexelene FX3-5N FX Flexible Polyethylene, Non-DEHP 3/16 x 5/16"" Tubing, 100 Ft
Catalog No. 50629616
Encompass
Applications:Food and beverage processing, instrumentation, pure-water transfer, water conditioning, Characteristics: Plasticizer-free, 78 A durometer, can be used with barbed fittings, abrasion-resistant, excellent chemical compatibility, economical. Colored Flexelene™ tubing available upon request. Certification: Produced with compounds listed by NSF International Standards 51 and 61, and are FDA compliant Temperature range:-40¿F to 170¿F (-40¿C to 76¿C) Sterilize: With ethylene oxide
Catalog No. 50-629-616 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9615504
Applications:Food and beverage processing, instrumentation, pure-water transfer, water conditioning, Characteristics: Plasticizer-free, 78 A durometer, can be used with barbed fittings, abrasion-resistant, excellent chemical compatibility, economical. Colored Flexelene™ tubing available upon request. Certification: Produced with compounds listed by NSF International Standards 51 and 61, and are FDA compliant Temperature range:-40¿F to 170¿F (-40¿C to 76¿C) Sterilize: With ethylene oxide