Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, High-Purity PFA-450, 1/16"" ID x 1/8"" OD; 25 ft
Catalog No. 50634344
Encompass
Applications: Pharmaceutical, biotechnology, environmental, semiconductor processing, food and beverage, autoanalyzer, and nuclear applications. Characteristics: Superior surface smoothness and clarity, low extractables with deionized water and acids, high resistance to stress and cracking. Translucent. Resin certification: Resin is FDA compliant. Tubing is manufactured according to GMP. Temperature range: -320 to 500¿F (-195 to 260¿C) Sterilize: With autoclave, ethylene oxide, gamma irradiation, and using dry heat.
Catalog No. 50-634-344 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636850
Applications: Pharmaceutical, biotechnology, environmental, semiconductor processing, food and beverage, autoanalyzer, and nuclear applications. Characteristics: Superior surface smoothness and clarity, low extractables with deionized water and acids, high resistance to stress and cracking. Translucent. Resin certification: Resin is FDA compliant. Tubing is manufactured according to GMP. Temperature range: -320 to 500¿F (-195 to 260¿C) Sterilize: With autoclave, ethylene oxide, gamma irradiation, and using dry heat.