Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 24"" Long; 5/PK
Catalog No. NC9551693
Encompass
This high pressure flexible PVC tubing has luer fittings at each end and comes in various lengths. Can be sterilized by ETO.
Catalog No. NC9551693 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3060060
This high pressure flexible PVC tubing has luer fittings at each end and comes in various lengths. Can be sterilized by ETO.