Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S High-Performance Precision Pump Tubing, Tygon Chemical, L/S 24; 50 ft
Catalog No. NC0202239
Encompass
Applications: Chemical, food, beverage, and pharmaceutical applications Characteristics: Excellent chemical resistance, low extractability, low gas permeability, plasticizer free, translucent NOTE: Tygon Chemical pump tubing is recommended for use in Easy-Load and Easy-Load II pump heads only.
Catalog No. NC0202239 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0647524/DEL
Applications: Chemical, food, beverage, and pharmaceutical applications Characteristics: Excellent chemical resistance, low extractability, low gas permeability, plasticizer free, translucent NOTE: Tygon Chemical pump tubing is recommended for use in Easy-Load and Easy-Load II pump heads only.