Learn More
IDEX Health & Science Upchurch TUBNG NAT 1/16ODX.040ID 50FT
Catalog No. NC1701216
Encompass_Preferred
PFA TUBING NATURAL 1/16"OD X .040"ID X 50FT; Material: Genuine Teflon PFA Resin, Pressure Rating Max PSI: 500; Pressure Rating Max Bar: 34; IDEX HEALTH & SCIENCE UPCHURCH mfg# 1507L
Catalog No. NC1701216 Supplier IDEX Health & Science Upchurch Supplier No. 1507L
PFA TUBING NATURAL 1/16"OD X .040"ID X 50FT; Material: Genuine Teflon PFA Resin, Pressure Rating Max PSI: 500; Pressure Rating Max Bar: 34; IDEX HEALTH & SCIENCE UPCHURCH mfg# 1507L