Learn More
IDEX Health & Science Upchurch PEEK Tubing 1/16" OD x .010" ID Blue 5ft
Catalog No. NC1071292
Encompass_Preferred
Biocompatible, PEEK Tubing is frequently used in high-pressure applications. Its distinct material delivers excellent chemical resistance and is specifically designed to be inert to most solvents. PEEK tubing offers a smooth internal surface that improves the resolution of sample bands. We have developed a variety of color options for easy coding and identification. It is available in lengths of 5ft, 50ft and 100ft. Colors available are blue and natural.
Catalog No. NC1071292 Supplier IDEX Health & Science Upchurch Supplier No. 1531B
Biocompatible, PEEK Tubing is frequently used in high-pressure applications. Its distinct material delivers excellent chemical resistance and is specifically designed to be inert to most solvents. PEEK tubing offers a smooth internal surface that improves the resolution of sample bands. We have developed a variety of color options for easy coding and identification. It is available in lengths of 5ft, 50ft and 100ft. Colors available are blue and natural.