Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, PFA, 1/16"" ID"" ID x 1/8"" OD; 25 ft
Catalog No. 50634307
Encompass
Ideal for general-purpose, single-use, and extended fluid path applications. Global market leader in range of formulations for most extensive chemical compatibility. Single-use custom lengths and configurations available to minimize downtime. Regulatory approvals to match your unique application.
Catalog No. 50-634-307 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0637501
Ideal for general-purpose, single-use, and extended fluid path applications. Global market leader in range of formulations for most extensive chemical compatibility. Single-use custom lengths and configurations available to minimize downtime. Regulatory approvals to match your unique application.