Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, PFA, 5/32""ID"" ID x 1/4"" OD; 25 ft
Catalog No. 50634290
Encompass
Zeus Industrial Products Inc model number 40257, PFA tubing, 5/32In ID x 1/4In OD, 25 ft/pack
Catalog No. 50-634-290 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0637502
Zeus Industrial Products Inc model number 40257, PFA tubing, 5/32In ID x 1/4In OD, 25 ft/pack