Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Microbore PTFE, 0.053"" ID x 0.076"" OD; 100 ft/roll
Catalog No. 50634311
Encompass
Use this translucent tubing in extreme temperatures and in harsh chemical environments. It's chemically inert, has low permeability, and has the lowest friction coefficient of any tubing. Temperature range is ¿400 to 500¿F (¿240 to 260¿C); melts at 620¿F (327¿C). Tensile strength is 2500 to 4000 psi. PTFE tubing resin is UL-recognized and complies with FDA (21 CFR 177.1550) and USP Class VI requirements.
Catalog No. 50-634-311 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0641751
Use this translucent tubing in extreme temperatures and in harsh chemical environments. It's chemically inert, has low permeability, and has the lowest friction coefficient of any tubing. Temperature range is ¿400 to 500¿F (¿240 to 260¿C); melts at 620¿F (327¿C). Tensile strength is 2500 to 4000 psi. PTFE tubing resin is UL-recognized and complies with FDA (21 CFR 177.1550) and USP Class VI requirements.