Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Peroxide-Cured PTFE, Blue, 1/16"" ID x 1/8"" OD; 12 ft
Catalog No. 50634297
Encompass
Applications: Ideal for dedicated or multiple lines¿provides quick and easy line identification. Characteristics: Chemically inert, nontoxic, and virtually nonporous. Opaque. Certifications: None. Temperature range: -400 to 500¿F (-240 to 260¿C). Sterilize: Autoclave, ethylene oxide, or dry heat.
Catalog No. 50-634-297 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9613022
Applications: Ideal for dedicated or multiple lines¿provides quick and easy line identification. Characteristics: Chemically inert, nontoxic, and virtually nonporous. Opaque. Certifications: None. Temperature range: -400 to 500¿F (-240 to 260¿C). Sterilize: Autoclave, ethylene oxide, or dry heat.