Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain Tygon Tubing, 1 in ID, 1.375 in OD

Catalog No. 502365835
Encompass_Preferred

Tygon 2475 is a crystal-clear, plasticizer-free thermoplastic tubing engineered for medical and diagnostic fluid transfer applications where low extractables and chemical compatibility are required. It provides a smooth inner bore to limit protein and particle retention and is suitable for cleanroom and analyzer fluid lines.

  • Plasticizer-free formulation to minimize leachables
  • 1.00 in (25.4 mm) ID, 1.375 in (34.9 mm) OD, 3/16 in (4.8 mm) wall
  • 25 ft (7.6 m) package length
  • USP Class VI biocompatibility
  • Brittleness temperature -108 °F (-78 °C) (ASTM D746)
  • Applications: diagnostic reagent delivery, medical dispensing, cell and tissue transport, cleanroom transfer lines

Catalog No. 50-236-5835 Supplier Accuflow Systems Supplier No. ACG42064
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tygon 2475 is a crystal-clear, plasticizer-free thermoplastic tubing engineered for medical and diagnostic fluid transfer applications where low extractables and chemical compatibility are required. It provides a smooth inner bore to limit protein and particle retention and is suitable for cleanroom and analyzer fluid lines.

  • Plasticizer-free formulation to minimize leachables
  • 1.00 in (25.4 mm) ID, 1.375 in (34.9 mm) OD, 3/16 in (4.8 mm) wall
  • 25 ft (7.6 m) package length
  • USP Class VI biocompatibility
  • Brittleness temperature -108 °F (-78 °C) (ASTM D746)
  • Applications: diagnostic reagent delivery, medical dispensing, cell and tissue transport, cleanroom transfer lines

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.