Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 2-Stop, Viton, 1.42 mm ID; 12/PK
Catalog No. NC1572560
Encompass
Applications: Acid and solvent transfer; high-temperature work Characteristics: Opaque black Resin certification: Tubing is manufactured according to GMP Temperature range: -8 to 400¿F (-13 to 204¿C) Sterilize: At 480¿F (249¿C) for 16 hours in a circulating hot air oven
The overall length of Viton 97628-series stop tubing is 12" (304 mm). The legacy Ismatec stop tubing was 180 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Catalog No. NC1572560 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9762834
Applications: Acid and solvent transfer; high-temperature work Characteristics: Opaque black Resin certification: Tubing is manufactured according to GMP Temperature range: -8 to 400¿F (-13 to 204¿C) Sterilize: At 480¿F (249¿C) for 16 hours in a circulating hot air oven
The overall length of Viton 97628-series stop tubing is 12" (304 mm). The legacy Ismatec stop tubing was 180 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.