Learn More
Medchemexpress LLC Tubulin cofactor A 50ug
SDP
Catalog No. 5000180132
Encompass_Preferred
The tubulin cofactor A protein is a key tubulin folding protein that initiates the tubulin folding pathway within the supercomplex (cofactors A to E) Cofactors A and D capture tubulin and stabilize it in a quasi-native state Tubulin cofactor A Protein Human (Tag Free) is the recombinant human-derived Tubulin cofactor A protein expressed by E coli with tag free
Catalog No. 50-001-80132 Supplier Medchemexpress LLC Supplier No. HYP73565A50UG
The tubulin cofactor A protein is a key tubulin folding protein that initiates the tubulin folding pathway within the supercomplex (cofactors A to E) Cofactors A and D capture tubulin and stabilize it in a quasi-native state Tubulin cofactor A Protein Human (Tag Free) is the recombinant human-derived Tubulin cofactor A protein expressed by E coli with tag free