Learn More
AdipoGen anti-Tubulin-GTP, mAb (rec.) (MB11) (100 µg)
SDP
Supplier:  AdipoGen AG27B0009C100
Encompass_Preferred
Recombinant Antibody. Recognizes human, mouse, rat and drosophila tubulin-GTP. Species cross-reactivity: Drosophila, Human, Mouse, Rat. Clone: MB11. Isotype: Human IgG2lambda. Applications: ICC. Host: Purified from HEK 293 cell culture supernatant. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. anti-Tubulin-GTP, monoclonal antibody (recombinant) (MB11) is an antibody developed by antibody phage display technology using a human naive antibody gene library.
Catalog No. 50-205-3026