Learn More
Medchemexpress LLC HY-112607 10mg Medchemexpress, Tubulin inhibitor 1 CAS:2237054-53-2 Purity:>98%
SDP
Catalog No. 501969112
Encompass_Preferred
Medchemexpress, HY-112607 10mg Tubulin inhibitor 1 CAS:2237054-53-2 Tubulin inhibitor 1 is a tubulin inhibitor, inhibits tubulin polymerization. Tubulin inhibitor 1 shows potent anti-tumor activity, casues cellular mitotic arrest in the G2/M phase, and induces cellular apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9112 Supplier Medchemexpress LLC Supplier No. HY11260710MG
Medchemexpress, HY-112607 10mg Tubulin inhibitor 1 CAS:2237054-53-2 Tubulin inhibitor 1 is a tubulin inhibitor, inhibits tubulin polymerization. Tubulin inhibitor 1 shows potent anti-tumor activity, casues cellular mitotic arrest in the G2/M phase, and induces cellular apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.