Learn More
Medchemexpress LLC HY-112607 5mg Medchemexpress, Tubulin inhibitor 1 CAS:2237054-53-2 Purity:>98%
SDP
Catalog No. 501932426
Encompass_Preferred
Medchemexpress, HY-112607 5mg Tubulin inhibitor 1 CAS:2237054-53-2 Tubulin inhibitor 1 is a tubulin inhibitor, inhibits tubulin polymerization. Tubulin inhibitor 1 shows potent anti-tumor activity, casues cellular mitotic arrest in the G2/M phase, and induces cellular apoptosis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2426 Supplier Medchemexpress LLC Supplier No. HY1126075MG
Medchemexpress, HY-112607 5mg Tubulin inhibitor 1 CAS:2237054-53-2 Tubulin inhibitor 1 is a tubulin inhibitor, inhibits tubulin polymerization. Tubulin inhibitor 1 shows potent anti-tumor activity, casues cellular mitotic arrest in the G2/M phase, and induces cellular apoptosis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.