Learn More
Medchemexpress LLC Tubulin inhibitor 11 | 2366260-33-3 | C22H23N3O3S | 1 ML
SDP
Catalog No. 5000373200
Encompass_Preferred
Tubulin inhibitor 11 is a potent and orally active tubulin inhibitor. It targets the Colchicine binding site on tubulin, inhibits tubulin polymerization, and promotes mitotic blockade and apoptosis.
  • Potent and orally active tubulin inhibitor.
  • Targets the colchicine binding site on tubulin.
  • Inhibits tubulin polymerization.
  • Promotes mitotic blockade and apoptosis.
  • Demonstrates antiproliferative activity against various human cancer cell lines and mouse B16-F10 cells.
  • Effective against paclitaxel-resistant cells.
  • Induces G2/M cell cycle arrest.
  • Shows robust in vivo antitumor efficacies.
Catalog No. 50-003-73200 Supplier Medchemexpress LLC Supplier No. HY15215610MM/1ML
Add to Cart
Add to Cart
Tubulin inhibitor 11 is a potent and orally active tubulin inhibitor. It targets the Colchicine binding site on tubulin, inhibits tubulin polymerization, and promotes mitotic blockade and apoptosis.
  • Potent and orally active tubulin inhibitor.
  • Targets the colchicine binding site on tubulin.
  • Inhibits tubulin polymerization.
  • Promotes mitotic blockade and apoptosis.
  • Demonstrates antiproliferative activity against various human cancer cell lines and mouse B16-F10 cells.
  • Effective against paclitaxel-resistant cells.
  • Induces G2/M cell cycle arrest.
  • Shows robust in vivo antitumor efficacies.