Learn More
Medchemexpress LLC Tubulin inhibitor 6 (compound 14b) | 105925-39-1 | 99.0% | C20H14ClNO2S | 10MG
SDP
Catalog No. 5000260945
Encompass_Preferred
Tubulin inhibitor 6 is a small-molecule tubulin polymerization inhibitor (Compound 14b) used for preclinical research. It has molecular formula C20H14ClNO2S, molecular weight 367.85 g/mol, CAS 105925-39-1, and shows activity against tubulin (IC50 = 0.87 μM) and K562 cell growth (IC50 = 840 nM).
  • Inhibits tubulin polymerization (IC50 = 0.87 μM).
  • Reduces K562 cell growth (IC50 = 840 nM).
  • High reported purity (99.0%).
  • Available in small-mass quantities suitable for screening and assay development.
  • Characterized by molecular formula C20H14ClNO2S and molecular weight 367.85 g/mol.
Catalog No. 50-002-60945 Supplier Medchemexpress LLC Supplier No. HY13612110MG
Tubulin inhibitor 6 is a small-molecule tubulin polymerization inhibitor (Compound 14b) used for preclinical research. It has molecular formula C20H14ClNO2S, molecular weight 367.85 g/mol, CAS 105925-39-1, and shows activity against tubulin (IC50 = 0.87 μM) and K562 cell growth (IC50 = 840 nM).
  • Inhibits tubulin polymerization (IC50 = 0.87 μM).
  • Reduces K562 cell growth (IC50 = 840 nM).
  • High reported purity (99.0%).
  • Available in small-mass quantities suitable for screening and assay development.
  • Characterized by molecular formula C20H14ClNO2S and molecular weight 367.85 g/mol.