Learn More
Medchemexpress LLC Tubuloside A | 112516-05-9 | 1 MG
SDP
Catalog No. 5000420626
Encompass_Preferred
Tubuloside A is a phenylethanoid glycoside with antioxidative effect and hepatoprotective activity.
  • Has antioxidative effect
  • Offers hepatoprotective activity
  • Inhibits D-GalN-induced death of hepatocytes
  • Appears as an off-white to light yellow solid
  • Derived from Cistanche deserticola Ma
  • Soluble in DMSO
Catalog No. 50-004-20626 Supplier Medchemexpress LLC Supplier No. HYN21551MG
Tubuloside A is a phenylethanoid glycoside with antioxidative effect and hepatoprotective activity.
  • Has antioxidative effect
  • Offers hepatoprotective activity
  • Inhibits D-GalN-induced death of hepatocytes
  • Appears as an off-white to light yellow solid
  • Derived from Cistanche deserticola Ma
  • Soluble in DMSO