Learn More
Medchemexpress LLC Tubulysin A | 205304-86-5 | 99.1% | 844.07 | C43H65N5O10S | 1 MG
SDP
Catalog No. 5000204247
Encompass_Preferred
Tubulysin A is a myxobacterial-derived cytotoxic research compound with anticancer and antiangiogenic activity. It acts as an anti-microtubule, anti-mitosis, and anti-proliferative agent and is commonly used in preclinical in vitro studies and as a payload in ADC research.
  • Anticancer and antiangiogenic activity.
  • Anti-microtubule and anti-mitosis mechanism.
  • High reported purity suitable for research use.
  • Available in small research quantities for preclinical studies.
  • Store protected from light and under inert atmosphere for stability.
Catalog No. 50-002-04247 Supplier Medchemexpress LLC Supplier No. HY159951MG
Tubulysin A is a myxobacterial-derived cytotoxic research compound with anticancer and antiangiogenic activity. It acts as an anti-microtubule, anti-mitosis, and anti-proliferative agent and is commonly used in preclinical in vitro studies and as a payload in ADC research.
  • Anticancer and antiangiogenic activity.
  • Anti-microtubule and anti-mitosis mechanism.
  • High reported purity suitable for research use.
  • Available in small research quantities for preclinical studies.
  • Store protected from light and under inert atmosphere for stability.