Learn More
Medchemexpress LLC Tubulysin M | 936691-46-2 | 1 MG
SDP
Catalog No. 5000419004
Encompass_Preferred
Tubulysin M is a highly cytotoxic anti-microtubule toxin, isolated from myxobacteria Archangium geophyra and Angiococcus disciformis. It is synthesized as an ADC cytotoxin and exhibits potent activity against various mammalian cells, including multidrug-resistant cell lines, with IC50 values in the low nanomolar range.
  • Inhibits microtubule/tubulin polymerization
  • Leads to cell cycle arrest and apoptosis
  • Active against multidrug-resistant cell lines
  • Binds to vinca domain and tubulin β2 subunit
Catalog No. 50-004-19004 Supplier Medchemexpress LLC Supplier No. HYN70531MG
Tubulysin M is a highly cytotoxic anti-microtubule toxin, isolated from myxobacteria Archangium geophyra and Angiococcus disciformis. It is synthesized as an ADC cytotoxin and exhibits potent activity against various mammalian cells, including multidrug-resistant cell lines, with IC50 values in the low nanomolar range.
  • Inhibits microtubule/tubulin polymerization
  • Leads to cell cycle arrest and apoptosis
  • Active against multidrug-resistant cell lines
  • Binds to vinca domain and tubulin β2 subunit