Learn More
Medchemexpress LLC HY-16069 50mg Medchemexpress, Tucatinib CAS:937263-43-9 Purity:>98%
SDP
Catalog No. 502029187
Encompass_Preferred
Medchemexpress, HY-16069 50mg Tucatinib CAS:937263-43-9 Tucatinib (Irbinitinib; ARRY-380; ONT-380) is a potent and selective HER2 inhibitor with an IC50 of 8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9187 Supplier Medchemexpress LLC Supplier No. HY1606950MG
Medchemexpress, HY-16069 50mg Tucatinib CAS:937263-43-9 Tucatinib (Irbinitinib; ARRY-380; ONT-380) is a potent and selective HER2 inhibitor with an IC50 of 8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.