Learn More
Medchemexpress LLC Tucatinib hemiethanolate | 1429755-56-5 | 100.0% | 503.57 g·mol⁻¹ | C26H24N8O2 · 1/2 C2H6O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16069A10MG
Encompass_Preferred
Tucatinib hemiethanolate is a research-grade small molecule that potently and selectively inhibits HER2 (reported IC50 8 nM). It is supplied as the hemiethanolate salt for analytical and research applications and is not for human therapeutic use.
  • Potent HER2 inhibition (IC50 8 nM).
  • Selective activity for HER2, supporting targeted research applications.
  • High purity suitable for analytical and in vitro studies.
  • Available in small research-scale quantities, such as 10 mg.
  • Provided with batch-specific certificate of analysis for traceability.
Catalog No. 50-002-85400