Learn More
Medchemexpress LLC Tucatinib hemiethanolate | 1429755-56-5 | 98.8% | 503.57 | 100 MG
SDP
Catalog No. 5000392871
Encompass_Preferred
Tucatinib hemiethanolate is a potent, orally active, and selective HER2 inhibitor. With an IC50 of 8 nM, it selectively targets HER2, showing approximately 500-fold selectivity over EGFR in cell-based assays. This compound is intended for research use only.
  • Potent, orally active, and selective HER2 inhibitor
  • Selectively inhibits receptor tyrosine kinase HER2 relative to EGFR
  • Blocks proliferation and phosphorylation of HER2 and its downstream effector Akt
  • Demonstrated anti-tumor activity and improved survival in animal models
Catalog No. 50-003-92871 Supplier Medchemexpress LLC Supplier No. HY16069A100MG
Tucatinib hemiethanolate is a potent, orally active, and selective HER2 inhibitor. With an IC50 of 8 nM, it selectively targets HER2, showing approximately 500-fold selectivity over EGFR in cell-based assays. This compound is intended for research use only.
  • Potent, orally active, and selective HER2 inhibitor
  • Selectively inhibits receptor tyrosine kinase HER2 relative to EGFR
  • Blocks proliferation and phosphorylation of HER2 and its downstream effector Akt
  • Demonstrated anti-tumor activity and improved survival in animal models