Learn More
Medchemexpress LLC Tucatinib hemiethanolate | 1429755-56-5 | 98.8% | 503.57 | 50 MG
SDP
Catalog No. 5000393086
Encompass_Preferred
Tucatinib hemiethanolate is a potent, orally active, and selective HER2 inhibitor. It demonstrates nanomolar activity against purified HER2 enzyme and is approximately 500-fold selective for HER2 versus EGFR in cell-based assays. It selectively inhibits the receptor tyrosine kinase HER2 relative to EGFR, blocking proliferation and phosphorylation of HER2 and its downstream effector, Akt in HER2 overexpressing cell lines. It is intended for research use only.
  • Potent and selective HER2 inhibition (IC50 of 8 nM)
  • Antiproliferative effects in HER2 overexpressing cell lines
  • Reduced EGFR-related toxicities
  • Anti-tumor activity and survival benefit in animal models
  • Significant reduction in brain pErbB2 (80%)
  • Dose-related tumor growth inhibition
  • Enhanced tumor growth inhibition when combined with trastuzumab
  • Studied in various clinical trials
Catalog No. 50-003-93086 Supplier Medchemexpress LLC Supplier No. HY16069A50MG
Tucatinib hemiethanolate is a potent, orally active, and selective HER2 inhibitor. It demonstrates nanomolar activity against purified HER2 enzyme and is approximately 500-fold selective for HER2 versus EGFR in cell-based assays. It selectively inhibits the receptor tyrosine kinase HER2 relative to EGFR, blocking proliferation and phosphorylation of HER2 and its downstream effector, Akt in HER2 overexpressing cell lines. It is intended for research use only.
  • Potent and selective HER2 inhibition (IC50 of 8 nM)
  • Antiproliferative effects in HER2 overexpressing cell lines
  • Reduced EGFR-related toxicities
  • Anti-tumor activity and survival benefit in animal models
  • Significant reduction in brain pErbB2 (80%)
  • Dose-related tumor growth inhibition
  • Enhanced tumor growth inhibition when combined with trastuzumab
  • Studied in various clinical trials