Learn More
Medchemexpress LLC HY-109015 10mg , Tucidinostat CAS:1616493-44-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10901510MG
Encompass_Preferred
HY-109015 10mg Tucidinostat CAS: 1616493-44-7 Tucidinostat (Chidamide) is a potent and orally bioavailable HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10) inhibitor, with IC50s of 95, 160, 67 and 78 nM, less active on HDAC8 and HDAC11 (IC50s, 733 nM, 432 nM, respectively), and shows no effect on HDAC4/5/6/7/9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9924