Learn More
TARGETMOL CHEMICALS INC TUCIDINOSTAT 25MG
SDP
Catalog No. 502579879
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tucidinostat (Chidamide) is an effective and orally bioavailable HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10) inhibitor with IC50s of 95 160 67 and 78 nM less active on HDAC8/11 (IC50 733/432 nM) and shows no effect on HDAC4/5/6/7/9. purity: 97%
Catalog No. 50-257-9879 Supplier TARGETMOL CHEMICALS INC Supplier No. T448125MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tucidinostat (Chidamide) is an effective and orally bioavailable HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10) inhibitor with IC50s of 95 160 67 and 78 nM less active on HDAC8/11 (IC50 733/432 nM) and shows no effect on HDAC4/5/6/7/9. purity: 97%