Learn More
Selleck Chemical LLC Tucidinostat (Chidamide) 5mg 1616493-44-7 HBI-8000, CS-055
SDP
Supplier:  Selleck Chemical LLC S85675MG
Encompass_Preferred
Tucidinostat (Chidamide, HBI-8000, CS-055) is a low nanomolar inhibitor of HDAC1, 2, 3, and 10, the HDAC isotypes well documented to be associated with the malignant phenotype with IC50 values of 95, 160, 67, 78 nM for HDAC1, 2, 3, 10 respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8068