Learn More
Medchemexpress LLC Tucidinostat-d4 | 1616493-44-7 | 98.6% | 394.43 | 10 MG
SDP
Catalog No. 5000345153
Encompass_Preferred
Tucidinostat-d4 is the deuterium labeled Tucidinostat. Tucidinostat is a potent and orally bioavailable inhibitor of HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10), with IC50s of 95, 160, 67 and 78 nM, respectively.
  • It can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-45153 Supplier Medchemexpress LLC Supplier No. HY109015S1MG
Tucidinostat-d4 is the deuterium labeled Tucidinostat. Tucidinostat is a potent and orally bioavailable inhibitor of HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10), with IC50s of 95, 160, 67 and 78 nM, respectively.
  • It can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.