Learn More
Fine Science Tools Mayo Scissors, ToughCut, Stainless Steel, 15 cm
SDP
Catalog No. NC9574449
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 39mm/ Alloy / Material: Stainless Steel/ Length: 15cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC9574449 Supplier Fine Science Tools Supplier No. 1411015
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 39mm/ Alloy / Material: Stainless Steel/ Length: 15cm/ Serrated: Yes/ Feature: ToughCut