Learn More
Fine Science Tools Surgical Scissors, ToughCut, Stainless Steel, 13 cm
SDP
Catalog No. NC9574451
Encompass
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 41mm/ Alloy / Material: Stainless Steel/ Length: 13cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC9574451 Supplier Fine Science Tools Supplier No. 1405413
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 41mm/ Alloy / Material: Stainless Steel/ Length: 13cm/ Serrated: Yes/ Feature: ToughCut