Learn More
MSC Lift-All Flat Eye and Eye Web Sling, Polyester, 6 in. Width, 20 ft. Length, 1 ea.
Catalog No. 5000515676
Encompass
This heavy-duty, two-ply polyester web sling features a flat eye and eye design, engineered for reliable lifting applications in material handling. The sling incorporates a tubular edge construction with a damage-resistant core to protect body fibers from cutting. It is rated for a vertical capacity of 16,500 lb, a choker capacity of 13,200 lb, and a basket capacity of 33,000 lb.
  • Features blue tubular edges and an edge damage limit (EDL) marker for quick identification
  • Includes a red core yarn warning system to visually assist with inspection
  • Equipped with an abrasion-resistant yellow yarn coating for extended service life
  • Complies with WSTDA, ASME B30.9, and OSHA 1910.184 standards
  • Operating temperature range from -60°F to 200°F
Catalog No. 50-005-15676 Supplier MSC Supplier No. 85820991
This heavy-duty, two-ply polyester web sling features a flat eye and eye design, engineered for reliable lifting applications in material handling. The sling incorporates a tubular edge construction with a damage-resistant core to protect body fibers from cutting. It is rated for a vertical capacity of 16,500 lb, a choker capacity of 13,200 lb, and a basket capacity of 33,000 lb.
  • Features blue tubular edges and an edge damage limit (EDL) marker for quick identification
  • Includes a red core yarn warning system to visually assist with inspection
  • Equipped with an abrasion-resistant yellow yarn coating for extended service life
  • Complies with WSTDA, ASME B30.9, and OSHA 1910.184 standards
  • Operating temperature range from -60°F to 200°F