Learn More
MSC TUFFMASTER CLR FACESHIELD
Catalog No. 5000788188
Encompass
This replacement window is designed for use with 3M headgear, specifically Aearo AOSafety 82500 & 82501 models. It features an easy-change design that mounts quickly inside the crown for added splash protection.
  • Molded stock construction
  • Bright/uncoated propionate material
  • No lens shade
  • Window measures 9 in high x 14-1/2 in wide
  • Thickness is 0.8 in
  • Applicable for welding, sanding, machine operations, cleaning, demolition, electrical, remediation, grinding, assembly, painting, and facility maintenance
  • Meets ANSI Z87.1 and CSA Z94.3 standards
Catalog No. 50-007-88188 Supplier MSC Supplier No. 09797226
This replacement window is designed for use with 3M headgear, specifically Aearo AOSafety 82500 & 82501 models. It features an easy-change design that mounts quickly inside the crown for added splash protection.
  • Molded stock construction
  • Bright/uncoated propionate material
  • No lens shade
  • Window measures 9 in high x 14-1/2 in wide
  • Thickness is 0.8 in
  • Applicable for welding, sanding, machine operations, cleaning, demolition, electrical, remediation, grinding, assembly, painting, and facility maintenance
  • Meets ANSI Z87.1 and CSA Z94.3 standards