Learn More
MSC TUFFMASTER FACESHLD
Catalog No. 5000787084
Encompass
This face shield window is made from polycarbonate and features a bright/uncoated finish. It provides protection for various applications, meeting both ANSI Z87.1 and CSA Z94.3 standards.
  • Made from polycarbonate material
  • Bright/uncoated coating
  • Green color
  • Features 0.08 inch thickness
  • Meets ANSI Z87.1 and CSA Z94.3 standards
  • Suitable for welding, sanding, machine operations, demolition, cleaning, remediation, electrical, grinding, assembly, painting, and facility maintenance
Catalog No. 50-007-87084 Supplier MSC Supplier No. 71273205
This face shield window is made from polycarbonate and features a bright/uncoated finish. It provides protection for various applications, meeting both ANSI Z87.1 and CSA Z94.3 standards.
  • Made from polycarbonate material
  • Bright/uncoated coating
  • Green color
  • Features 0.08 inch thickness
  • Meets ANSI Z87.1 and CSA Z94.3 standards
  • Suitable for welding, sanding, machine operations, demolition, cleaning, remediation, electrical, grinding, assembly, painting, and facility maintenance