Learn More
MSC Contec Professional TuffStuff Heavy Duty Wipes, Cellulose/Polyester Blend, 12 x 16.5 in, Double Pop-Up Box
Catalog No. 502838384
Encompass
These heavy-duty nonwoven wipes are designed for critical cleaning tasks in professional applications. Made from a hydroentangled cellulose/polyester fabric blend, they are solvent-resistant and maintain strength when wet or dry. They offer excellent absorption and are considered low lint, making them suitable for a variety of cleaning tasks, including absorbing spills, oils, and grime, and applying solvents.
  • Made from a durable, solvent-resistant cellulose/polyester blend that maintains strength when wet.
  • Nonwoven, creped construction with no binders or adhesives.
  • Considered low lint for critical cleaning tasks.
  • Offers excellent absorption for spills, oils, grime, and solvents.
  • Suitable for general purpose cleaning, machine part wiping, and applying solvents.
  • Available in various sizes and packaging configurations for flexible use.
Catalog No. 50-283-8384 Supplier MSC Supplier No. 35620848
These heavy-duty nonwoven wipes are designed for critical cleaning tasks in professional applications. Made from a hydroentangled cellulose/polyester fabric blend, they are solvent-resistant and maintain strength when wet or dry. They offer excellent absorption and are considered low lint, making them suitable for a variety of cleaning tasks, including absorbing spills, oils, and grime, and applying solvents.
  • Made from a durable, solvent-resistant cellulose/polyester blend that maintains strength when wet.
  • Nonwoven, creped construction with no binders or adhesives.
  • Considered low lint for critical cleaning tasks.
  • Offers excellent absorption for spills, oils, grime, and solvents.
  • Suitable for general purpose cleaning, machine part wiping, and applying solvents.
  • Available in various sizes and packaging configurations for flexible use.