Learn More
MSC Ansell ActivArmr Arm Sleeve, Kevlar, Knitwrist, 26 in., 12/Pk.
Catalog No. 5000688273
Encompass
These medium-duty protective arm sleeves are designed to provide moderate heat, cut, and abrasion resistance for enhanced safety in industrial environments. Composed of DuPont™ Kevlar® interlock fabric, these sleeves promote natural mobility and breathability, ensuring sustained comfort during daily use. The integrated knitwrist cuff style provides a secure fit that helps protect the wrists and prevents the ingress of dirt and debris, making them suitable for handling sharp or rough materials.
  • Moderate heat, cut, and abrasion resistance
  • Lightweight and breathable design for everyday comfort
  • Knitwrist cuff style for a secure fit
  • Cut and sewn construction
  • Ideal for riveting, welding, bonding, and handling metal sheets or sharp-edged components
Catalog No. 50-006-88273 Supplier MSC Supplier No. 89919062
These medium-duty protective arm sleeves are designed to provide moderate heat, cut, and abrasion resistance for enhanced safety in industrial environments. Composed of DuPont™ Kevlar® interlock fabric, these sleeves promote natural mobility and breathability, ensuring sustained comfort during daily use. The integrated knitwrist cuff style provides a secure fit that helps protect the wrists and prevents the ingress of dirt and debris, making them suitable for handling sharp or rough materials.
  • Moderate heat, cut, and abrasion resistance
  • Lightweight and breathable design for everyday comfort
  • Knitwrist cuff style for a secure fit
  • Cut and sewn construction
  • Ideal for riveting, welding, bonding, and handling metal sheets or sharp-edged components