Learn More
MSC Lift-All Endless Roundsling, Polyester, 12 ft, 13,200 lb Capacity, 1 ea
Catalog No. 5000510343
Encompass
This versatile endless Type 5 roundsling provides a 12 ft. length for various lifting applications. Constructed from polyester, the sling is designed for durability and ease of use in demanding environments. It features a color-coded capacity system for quick identification and is engineered to meet or exceed OSHA and ASME B30.9 standards.
  • Individual slings can be attached together using appropriate hardware
  • Independent woven jacket allows for wear points to be shifted, minimizing abrasive wear
  • Load stability and balance can be achieved by spreading sling legs
  • Most flexible style of sling
  • Abrasion-resistant design with no strength loss from abrasion to the cover
Catalog No. 50-005-10343 Supplier MSC Supplier No. 06273262
This versatile endless Type 5 roundsling provides a 12 ft. length for various lifting applications. Constructed from polyester, the sling is designed for durability and ease of use in demanding environments. It features a color-coded capacity system for quick identification and is engineered to meet or exceed OSHA and ASME B30.9 standards.
  • Individual slings can be attached together using appropriate hardware
  • Independent woven jacket allows for wear points to be shifted, minimizing abrasive wear
  • Load stability and balance can be achieved by spreading sling legs
  • Most flexible style of sling
  • Abrasion-resistant design with no strength loss from abrasion to the cover