Learn More
MSC Lift-All Tuflex Polyester Endless Round Sling, 10 ft., 2,600 lbs. Capacity, 1 ea.
Catalog No. 5000515964
Encompass
This endless synthetic sling is constructed from polyester yarn protected by a double-wall tubular jacket. The design allows internal core yarns to move independently, ensuring that core yarns carry 100% of the load while the jacket provides essential abrasion resistance. This lightweight and flexible sling is engineered for versatile rigging and extended service life.
  • Meets or exceeds OSHA and ASME B30.9 standards
  • Features double-wall protection for enhanced durability
  • Constructed with independent core yarn movement to reduce internal friction
  • Designed for easy rigging with high flexibility
  • Jacket serves as a visual wear indicator when core yarns become visible
  • Maximum operating temperature of 200°F
  • Vertical capacity of 2,600 lbs.
Catalog No. 50-005-15964 Supplier MSC Supplier No. 89673610
This endless synthetic sling is constructed from polyester yarn protected by a double-wall tubular jacket. The design allows internal core yarns to move independently, ensuring that core yarns carry 100% of the load while the jacket provides essential abrasion resistance. This lightweight and flexible sling is engineered for versatile rigging and extended service life.
  • Meets or exceeds OSHA and ASME B30.9 standards
  • Features double-wall protection for enhanced durability
  • Constructed with independent core yarn movement to reduce internal friction
  • Designed for easy rigging with high flexibility
  • Jacket serves as a visual wear indicator when core yarns become visible
  • Maximum operating temperature of 200°F
  • Vertical capacity of 2,600 lbs.