Learn More
MSC Lift-All Tuflex Polyester Endless Roundsling, 21,200 Lb. Capacity, 10 ft., 1 ea.
Catalog No. 5000515709
Encompass
This endless synthetic roundsling is constructed from polyester yarn protected by a double-wall tubular jacket. Designed for versatility and load stability, the core yarns move independently within the jacket to accommodate various rigging configurations, and the design allows for shifting wear points to extend the service life of the sling.
  • Features a Tufhide™ nylon jacket for enhanced abrasion resistance
  • Achieves load stability and balance by spreading sling legs
  • Compatible with appropriate hardware for attaching individual slings together
  • Operates at temperatures up to 200°F
  • Complies with OSHA and ASME B30.9 standards
  • Vertical rated capacity of 21,200 lbs.
Catalog No. 50-005-15709 Supplier MSC Supplier No. 89673974
This endless synthetic roundsling is constructed from polyester yarn protected by a double-wall tubular jacket. Designed for versatility and load stability, the core yarns move independently within the jacket to accommodate various rigging configurations, and the design allows for shifting wear points to extend the service life of the sling.
  • Features a Tufhide™ nylon jacket for enhanced abrasion resistance
  • Achieves load stability and balance by spreading sling legs
  • Compatible with appropriate hardware for attaching individual slings together
  • Operates at temperatures up to 200°F
  • Complies with OSHA and ASME B30.9 standards
  • Vertical rated capacity of 21,200 lbs.