Learn More
MSC Lift-All Tuflex Endless Round Sling, Polyester, 3 ft., 5,300 lb. Vertical Capacity, 1 ea.
Catalog No. 5000511426
Encompass
The Lift-All® Tuflex® endless round sling is a durable lifting solution designed for industrial applications. Featuring a continuous loop of polyester yarn protected by a double-wall tubular jacket, this sling provides exceptional strength and flexibility while conforming to various load shapes. It is color-coded for quick capacity identification and includes a permanent Tuff-Tag for OSHA-compliant traceability.
  • Provides a 5,300 lb. vertical capacity and 4,200 lb. choker capacity.
  • Features low stretch of approximately 3% at rated capacity for improved load control.
  • Constructed from polyester to resist UV degradation, grease, and most acids.
  • Operating temperature range of -50°F to 200°F.
  • Designed to meet or exceed OSHA and ASME B30.9 standards.
  • Lightweight and pliable design enhances portability and safety during rigging.
Catalog No. 50-005-11426 Supplier MSC Supplier No. 89751218
The Lift-All® Tuflex® endless round sling is a durable lifting solution designed for industrial applications. Featuring a continuous loop of polyester yarn protected by a double-wall tubular jacket, this sling provides exceptional strength and flexibility while conforming to various load shapes. It is color-coded for quick capacity identification and includes a permanent Tuff-Tag for OSHA-compliant traceability.
  • Provides a 5,300 lb. vertical capacity and 4,200 lb. choker capacity.
  • Features low stretch of approximately 3% at rated capacity for improved load control.
  • Constructed from polyester to resist UV degradation, grease, and most acids.
  • Operating temperature range of -50°F to 200°F.
  • Designed to meet or exceed OSHA and ASME B30.9 standards.
  • Lightweight and pliable design enhances portability and safety during rigging.