Learn More
Accuflow Systems TuFlux SIL 1/4"x7/16" (1/4 x 7/16 inch, 6,4 x 11,1 mm)
Catalog No. NC2187009
Encompass_Preferred
Manufactured by Sartorius, FSA117427 Tuflux SIL ¼’’ x 7/16’’, 6.4 x 11.1 mm wall thickness 2.4 mm, 1 coil = 50 m
This item is not returnable. View return policy
Catalog No. NC2187009 Supplier Accuflow Systems Supplier No. FSA117427
Manufactured by Sartorius, FSA117427 Tuflux SIL ¼’’ x 7/16’’, 6.4 x 11.1 mm wall thickness 2.4 mm, 1 coil = 50 m