Learn More
Accuflow Systems TuFlux SIL 3/8"x5/8" (3/8 x 5/8 inch, 9,6 x 15,9 mm)
Catalog No. NC2187010
Encompass_Preferred
Manufactured by Sartorius, FSA117428 Tuflux SIL ⅜’’ x ⅝’’, 9.5 x 15.9 mm wall thickness 3.2 mm, 1 coil = 25 m Piece $292 1 $292 1 piece
This item is not returnable. View return policy
Catalog No. NC2187010 Supplier Accuflow Systems Supplier No. FSA117428
Manufactured by Sartorius, FSA117428 Tuflux SIL ⅜’’ x ⅝’’, 9.5 x 15.9 mm wall thickness 3.2 mm, 1 coil = 25 m Piece $292 1 $292 1 piece