Learn More
Accuflow Systems TuFlux SIL 1/8"x1/4" (1/8 x 1/4 inch, 3,2 x 6,4 mm)
Catalog No. NC2187005
Encompass_Preferred
Manufactured by Sartorius, FSA117425 Tuflux SIL ⅛” x ¼”, 3.2 x 6.4 mm wall thickness 1.6 mm, 1 coil = 100 m Piece $571 1 $571 1 piece
This item is not returnable. View return policy
Catalog No. NC2187005 Supplier Accuflow Systems Supplier No. FSA117425
Manufactured by Sartorius, FSA117425 Tuflux SIL ⅛” x ¼”, 3.2 x 6.4 mm wall thickness 1.6 mm, 1 coil = 100 m Piece $571 1 $571 1 piece