Learn More
Accuflow Systems TuFlux SIL 1/2"x3/4" (1/2 x 3/4 inch, 12,7 x 19,1 mm)
Catalog No. NC2187011
Encompass_Preferred
Manufactured by Sartorius, FSA117429 Tuflux SIL ½’’ x ¾’’, 12.7 x 19.1 mm wall thickness 3.2 mm, 1 coil = 20 m Piece $439 1 $439 1 piece
This item is not returnable. View return policy
Catalog No. NC2187011 Supplier Accuflow Systems Supplier No. FSA117429
Manufactured by Sartorius, FSA117429 Tuflux SIL ½’’ x ¾’’, 12.7 x 19.1 mm wall thickness 3.2 mm, 1 coil = 20 m Piece $439 1 $439 1 piece