Learn More
Accuflow Systems TuFlux TPE 1/8"x1/4" (1/8 x 1/4 inch, 3,2 x 6,4 mm)
Catalog No. NC2186997
Encompass_Preferred
Manufactured by Sartorius, TuFlux TPE ⅛” x ¼” (3.2 x 6.4 mm x wall thickness 1.6 mm) 1 coil = 200 m
This item is not returnable. View return policy
Catalog No. NC2186997 Supplier Accuflow Systems Supplier No. FSA121869
Manufactured by Sartorius, TuFlux TPE ⅛” x ¼” (3.2 x 6.4 mm x wall thickness 1.6 mm) 1 coil = 200 m