Learn More
Tufpak Secura'T™ Autoclave Bags
Catalog No. 14375212
Change view
Click to view available options
Dimensions (L x W):
19 x 24
25 x 35
8 x 12
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
14-375-212 25 x 35
14-375-210 8 x 12
14-375-211 19 x 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-375-212 Supplier Tufpak Supplier No. 11122535 PIO
Clear film, printed bag with temperature indicator patch.
  • Made from SUPROP™ Polypropylene film, specifically engineered for processing biohazardous materials and waste products in an autoclave prior to disposal
  • Recommended for maximum temperature exposure to 140°C (285°F)
  • Twist ties are included to allow proper closure for normal bag expansion and steam venting during sterilization cycle
  • Bag exceeds strength requirements of 165g dart drop standard as performed under ASTM D1709-04
Specifications
Color Clear
Material Polypropylene Film
Dimensions (L x W) 25 x 35